Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021267.4 | NP_067090.1 | 8 | 2452 |
Gene Symbol: GDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001492.5 | NP_001483.3 | 8 | 2452 | ||
GenBank mRNA | M62302.1 | - | 8 | 2404 |