Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000174.4 | NP_000165.1 | 3 | 865 | ||
XM_005247374.3 | XP_005247431.1 | 5 | 1161 | |||
XM_011512701.1 | XP_011511003.1 | 6 | 1243 | |||
XM_011512702.1 | XP_011511004.1 | 5 | 1118 | |||
GenBank mRNA | BC030229.1 | - | 3 | 830 | 5226377 | |
X52997.1 | - | 3 | 862 |