Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004489.4 | NP_004480.1 | 4 | 438 | ||
GenBank mRNA | AK301877.1 | - | 5 | 450 | ||
BC013652.1 | - | 4 | 438 | |||
BC103901.1 | - | 5 | 663 | 40005162 | ||
BC103902.1 | - | 4 | 1024 | |||
BC103903.1 | - | 5 | 663 | 40005170 | ||
BC107738.1 | - | 5 | 473 | 5810755 | ||
BT006998.1 | - | 4 | 362 | |||
CR541723.1 | - | 4 | 362 | |||
CR541750.1 | - | 4 | 362 | |||
U28963.1 | - | 5 | 452 |