Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000850.4 | NP_000841.1 | 7 | 789 | ||
NM_147148.2 | NP_671489.1 | 7 | 789 | |||
NR_024538.1 | 6 | 707 | ||||
XM_017001085.1 | XP_016856574.1 | 7 | 789 | |||
XM_017001086.1 | XP_016856575.1 | 7 | 1021 | |||
XM_017001087.1 | XP_016856576.1 | 7 | 1015 | |||
XM_017001088.1 | XP_016856577.1 | 6 | 700 | |||
GenBank mRNA | AK291880.1 | - | 7 | 789 | ||
BC015513.1 | - | 7 | 749 | 3893061 | ||
BC108729.1 | - | 7 | 746 | 6171894 | ||
BI458242.1 | - | 7 | 790 | |||
CR541869.1 | - | 7 | 475 | |||
DQ062813.1 | - | 5 | 577 | |||
M96234.1 | - | 7 | 738 | |||
M99421.1 | - | 6 | 531 | |||
M99422.1 | - | 7 | 747 |