Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000558.4 | NP_000549.1 | 3 | 478 | ||
GenBank mRNA | AF097635.1 | - | 3 | 435 | ||
AF105974.1 | - | 3 | 433 | |||
AF147332.1 | - | 3 | 430 | |||
AF281258.1 | - | 2 | 260 | |||
AF349571.1 | - | 3 | 437 | |||
AI064720.1 | - | 2 | 236 | |||
AK223392.1 | - | 3 | 455 | |||
BC005931.1 | - | 3 | 449 | 4047053 | ||
BC008572.1 | - | 3 | 435 | 4177243 | ||
BC032122.2 | - | 3 | 435 | 4849981 | ||
BC050661.1 | - | 3 | 435 | 5209215 | ||
BC101846.1 | - | 3 | 442 | 8069352 | ||
BC101848.1 | - | 3 | 442 | 8069354 | ||
DQ655926.2 | - | 2 | 256 | |||
V00493.1 | - | 3 | 449 |
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 3 | 478 |