Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000184.2 | NP_000175.1 | 1, 2 | 154 | ||
GenBank mRNA | AF110493.1 | - | 1, 2 | 48 | ||
AK290492.1 | - | 1, 2 | 154 | |||
BC010914.1 | - | 1, 2 | 158 | 4134713 | ||
BC029387.1 | - | 1, 2 | 184 | 4762402 | ||
BC130457.1 | - | 1, 2 | 149 | 40146488 | ||
BC130459.1 | - | 1, 2 | 149 | 40146490 | ||
M15386.1 | - | 1, 2 | 118 | |||
X55656.1 | - | 1, 2 | 81 |
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 1, 2 | 154 | ||
GenBank mRNA | BC010913.1 | - | 1, 2 | 160 | 4133117 | |
BC020719.1 | - | 1, 2 | 154 | 4732536 |