Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002113.2 | NP_002104.2 | 4 | 589 | ||
XM_011509457.2 | XP_011507759.1 | 3 | 395 | |||
GenBank mRNA | BC016755.1 | - | 4 | 567 | 3934474 | |
BC107771.1 | - | 4 | 585 | 4712577 | ||
BG568702.1 | - | 4 | 586 | |||
M65293.1 | - | 4 | 550 | |||
X56210.1 | - | 4 | 515 |
Gene Symbol: CFHR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG618529.1 | - | 3 | 389 |