Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001540.3 | NP_001531.1 | 3 | 770 | ||
GenBank mRNA | AB020027.1 | - | 3 | 640 | ||
AK296890.1 | - | 4 | 550 | |||
AK311446.1 | - | 3 | 402 | |||
AL050380.1 | - | 4 | 501 | |||
BC000510.2 | - | 3 | 714 | 2822325 | ||
BC012292.1 | - | 3 | 688 | 3862626 | ||
BC012768.2 | - | 3 | 637 | 3689220 | ||
BC014920.1 | - | 3 | 574 | |||
BC073768.1 | - | 3 | 635 | 5444848 | ||
FJ224323.1 | - | 3 | 722 | |||
S74571.1 | - | 2 | 250 | |||
U90906.1 | - | 3 | 722 | |||
X16477.1 | - | 3 | 289 | |||
X54079.1 | - | 3 | 667 |