Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 644 | ||
GenBank mRNA | BC113994.1 | - | 1 | 644 | 40084821 | |
BC114000.1 | - | 1 | 644 | 40084820 | ||
M34913.1 | - | 1 | 644 |