Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001035235.3 | NP_001030312.2 | 5 | 1198 | ||
NM_001253764.1 | NP_001240693.1 | 3 | 513 | |||
NR_045586.1 | 5 | 1258 | ||||
NR_045587.1 | 4 | 716 | ||||
GenBank mRNA | AF092038.1 | - | 5 | 560 | ||
AF293024.1 | - | LIT1246 SRA |
5 | 782 | ||
AF293025.1 | - | LIT1250 SRA |
5 | 782 | ||
AF293026.1 | - | LIT1251 SRA |
5 | 785 | ||
AF318361.1 | - | 5 | 1202 | |||
AK024640.1 | - | 4 | 702 | |||
AK054960.1 | - | 5 | 1418 | |||
BC040043.2 | - | 4 | 530 | |||
BU553214.1 | - | 3 | 533 | |||
DQ286291.1 | - | 5 | 842 |