Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004981.1 | NP_004972.1 | 2 | 188 | ||
NM_152868.2 | NP_690607.1 | 2 | 350 | |||
GenBank mRNA | BC032562.1 | - | 2 | 360 | ||
BC039618.1 | - | 2 | 233 | |||
BC113506.1 | - | 1 | 142 | 8322558 | ||
BC113508.1 | - | 1 | 142 | 8322560 | ||
BM475496.1 | - | 348 | ||||
CR456507.1 | - | 2 | 158 | |||
S72503.1 | - | 2 | 189 | |||
U07364.1 | - | 2 | 188 | |||
U24056.1 | - | 2 | 325 |