Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001035235.3 | NP_001030312.2 | 4 | 1065 | ||
NM_001253764.1 | NP_001240693.1 | 2 | 380 | |||
NR_045586.1 | 4 | 1125 | ||||
NR_045587.1 | 3 | 583 | ||||
GenBank mRNA | AF092038.1 | - | 4 | 427 | ||
AF293024.1 | - | LIT1246 SRA |
4 | 649 | ||
AF293025.1 | - | LIT1250 SRA |
4 | 649 | ||
AF293026.1 | - | LIT1251 SRA |
4 | 652 | ||
AF318361.1 | - | 4 | 1069 | |||
AK024640.1 | - | 3 | 569 | |||
AK054960.1 | - | 4 | 1285 | |||
BC040043.2 | - | 3 | 397 | |||
BU553214.1 | - | 2 | 400 | |||
CX165918.1 | - | 4 | 687 |