Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005954.2 | NP_005945.1 | 3 | 553 | ||
GenBank mRNA | BC013081.1 | - | 3 | 364 | 4152899 | |
BI752092.1 | - | 541 | ||||
BI820409.1 | - | 555 | ||||
CB155600.1 | - | 3 | 411 | |||
D13365.1 | - | 3 | 368 |