Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINOS1-NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204088.1 | NP_001191017.1 | 3 | 410 | ||
NM_001204089.1 | NP_001191018.1 | 2 | 225 | |||
GenBank mRNA | AK289456.1 | - | 2 | 223 | ||
FY212253.1 | - | 3 | 319 |
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204084.2 | NP_001191013.1 | 2 | 333 | ||
NM_001204085.1 | NP_001191014.1 | 2 | 251 | |||
NM_001204086.2 | NP_001191015.1 | 2 | 360 | |||
NM_001278164.1 | NP_001265093.1 | 2 | 242 | |||
NM_001278165.1 | NP_001265094.1 | 2 | 309 | |||
NM_001278166.1 | NP_001265095.1 | 2 | 153 | |||
NM_005380.7 | NP_005371.2 | 2 | 366 | |||
NM_182744.3 | NP_877421.2 | 2 | 261 | |||
GenBank mRNA | AK292101.1 | - | 2 | 292 | ||
AK300872.1 | - | 2 | 261 | |||
AK309257.1 | - | 2 | 360 | |||
AK313265.1 | - | 2 | 251 | |||
AL703187.1 | - | 2 | 259 | |||
AY049783.1 | - | 1 | 90 | |||
BC012037.1 | - | 2 | 162 | 3874204 | ||
BQ671197.1 | - | 218 | ||||
BT019423.1 | - | 1 | 70 | |||
D28124.1 | - | 2 | 131 | |||
DA384331.1 | - | 2 | 309 | |||
DA921297.1 | - | 2 | 258 | |||
DA924824.1 | - | 2 | 333 | |||
DB016315.1 | - | 2 | 240 | |||
DB253522.1 | - | 2 | 261 | |||
DB482000.1 | - | 2 | 226 | |||
HY084536.1 | - | 2 | 149 |