Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002619.3 | NP_002610.1 | 3 | 500 | ||
XM_005265696.3 | XP_005265753.1 | 3 | 401 | |||
GenBank mRNA | AJ705588.1 | - | 3 | 374 | ||
M25897.1 | - | 3 | 336 |