Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002675.3 | NP_002666.1 | 3 | 892 | ||
NM_033238.2 | NP_150241.2 | 3 | 892 | |||
NM_033239.2 | NP_150242.1 | 3 | 892 | |||
NM_033240.2 | NP_150243.2 | 3 | 892 | |||
NM_033244.3 | NP_150247.2 | 3 | 892 | |||
NM_033246.2 | NP_150249.1 | 3 | 892 | |||
NM_033247.2 | NP_150250.2 | 3 | 892 | |||
NM_033249.2 | NP_150252.1 | 3 | 892 | |||
NM_033250.2 | NP_150253.2 | 3 | 892 | |||
GenBank mRNA | AB208950.1 | - | 3 | 762 | ||
AB209051.1 | - | 2 | 603 | |||
AB209411.1 | - | 3 | 836 | |||
AF230401.1 | - | 3 | 824 | |||
AF230402.1 | - | 3 | 824 | |||
AF230403.1 | - | 3 | 824 | |||
AF230404.1 | - | 3 | 824 | |||
AF230405.1 | - | 3 | 824 | |||
AF230406.1 | - | 3 | 824 | |||
AF230407.1 | - | 3 | 824 | |||
AF230408.1 | - | 3 | 824 | |||
AF230409.1 | - | 3 | 824 | |||
AF230410.1 | - | 3 | 824 | |||
AF230411.1 | - | 3 | 824 | |||
BC000080.2 | - | 3 | 813 | 3506984 | ||
BC020994.2 | - | 3 | 813 | 2958462 | ||
BT009911.1 | - | 3 | 752 | |||
M73778.1 | - | 3 | 832 | |||
M79462.1 | - | 3 | 766 | |||
M79463.1 | - | 3 | 766 | |||
M79464.1 | - | 3 | 766 | |||
M80185.1 | - | 3 | 766 | |||
S50913.1 | - | 3 | 834 | |||
X63131.1 | - | 3 | 893 |