Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002801.3 | NP_002792.1 | 3 | 300 | ||
GenBank mRNA | AK225658.1 | - | 3 | 273 | ||
AK298012.1 | - | 2 | 497 | |||
AK312208.1 | - | 3 | 260 | |||
BC017198.1 | - | 3 | 251 | 3143189 | ||
BC052369.1 | - | 3 | 238 | 4054165 | ||
BM845639.1 | - | 3 | 273 | |||
BP289065.1 | - | 3 | 300 | |||
BT019723.1 | - | 3 | 172 | |||
BT019724.1 | - | 3 | 172 | |||
CR456982.1 | - | 3 | 172 | |||
Y13640.1 | - | 3 | 260 |