Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000979.3 | NP_000970.1 | 3, 4 | 476 | ||
NM_001270490.1 | NP_001257419.1 | 2, 3 | 389 | |||
NR_073022.1 | 3, 4 | 468 | ||||
GenBank mRNA | AK026558.1 | - | 3, 4 | 234 | ||
AK293386.1 | - | 3, 4 | 235 | |||
BC000374.2 | - | 2, 3 | 208 | 2820119 | ||
BC009708.2 | - | 3, 4 | 222 | 3878799 | ||
BC021743.2 | - | 3, 4 | 234 | 5261251 | ||
BM764512.1 | - | 3, 4 | 594 | |||
CD246847.1 | - | 2, 3 | 173 | |||
CD389415.1 | - | 3, 4 | 240 | |||
DQ403032.1 | - | 2, 3 | 132 | |||
L11566.1 | - | 3, 4 | 216 |