Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000992.2 | NP_000983.1 | 4 | 224 | ||
GenBank mRNA | AK289538.1 | - | 4 | 177 | ||
AK311884.1 | - | 4 | 177 | |||
BC008926.2 | - | 4 | 224 | 3835176 | ||
BC070190.1 | - | 1 | 175 | 6452382 | ||
BC070481.1 | - | 4 | 163 | 6025704 | ||
BC071663.1 | - | 1 | 149 | 5220693 | ||
BC071909.1 | - | 4 | 162 | 6729601 | ||
U10248.1 | - | 4 | 159 | |||
U49083.1 | - | 4 | 157 | |||
Z49148.1 | - | 4 | 159 |
Gene Symbol: RPL29P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002778.1 | 1 | 197 | |||
GenBank mRNA | AF085886.1 | - | 1 | 197 |