Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291424.1 | NP_001278353.1 | 7 | 1071 | ||
NM_005475.2 | NP_005466.1 | 8 | 1994 | |||
XM_005253818.4 | XP_005253875.1 | 9 | 2138 | |||
XM_005253819.4 | XP_005253876.1 | 8 | 2021 | |||
XM_006719180.3 | XP_006719243.1 | 7 | 1623 | |||
XM_011537719.2 | XP_011536021.1 | 9 | 2141 | |||
XM_011537720.2 | XP_011536022.1 | 9 | 1806 | |||
XM_011537721.2 | XP_011536023.1 | 7 | 3606 | |||
GenBank mRNA | AB208911.1 | - | 7 | 1230 | ||
AF055581.1 | - | 8 | 1994 | |||
AJ012793.1 | - | 7 | 1640 | |||
AK302173.1 | - | 7 | 1071 | |||
BC136451.1 | - | 8 | 1680 | 9020440 |