Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291424.1 | NP_001278353.1 | 4 | 393 | ||
NM_005475.2 | NP_005466.1 | 5 | 1316 | |||
XM_005253818.4 | XP_005253875.1 | 6 | 1460 | |||
XM_005253819.4 | XP_005253876.1 | 5 | 1343 | |||
XM_006719180.3 | XP_006719243.1 | 4 | 945 | |||
XM_011537719.2 | XP_011536021.1 | 6 | 1463 | |||
XM_011537720.2 | XP_011536022.1 | 6 | 1128 | |||
XM_011537721.2 | XP_011536023.1 | 4 | 2928 | |||
GenBank mRNA | AB208911.1 | - | 4 | 552 | ||
AF055581.1 | - | 5 | 1316 | |||
AJ012793.1 | - | 4 | 962 | |||
AK302173.1 | - | 4 | 393 | |||
BC136451.1 | - | 5 | 1002 | 9020440 |