Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001022.3 | NP_001013.1 | 6 | 822 | ||
NM_001321483.1 | NP_001308412.1 | 6 | 499 | |||
NM_001321484.1 | NP_001308413.1 | 6 | 629 | |||
NM_001321485.1 | NP_001308414.1 | 6 | 835 | |||
GenBank mRNA | AA165492.1 | - | 4 | 377 | ||
BC000023.2 | - | 6 | 519 | 3504083 | ||
BC007615.2 | - | 5 | 450 | 3354280 | ||
BC017386.1 | - | 6 | 488 | |||
BC018616.1 | - | 6 | 822 | 4867036 | ||
BG398397.1 | - | 6 | 629 | |||
BU603942.1 | - | 6 | 501 | |||
CB124389.1 | - | 6 | 528 | |||
CB125125.1 | - | 6 | 499 | |||
M81757.1 | - | 6 | 472 |