Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024.3 | NP_001015.1 | 3 | 163 | ||
GenBank mRNA | AV758927.1 | - | 124 | |||
BC018140.1 | - | 2 | 100 | 3903320 | ||
BC021990.1 | - | 3 | 128 | 4066330 | ||
BM840443.1 | - | 3 | 163 | |||
CF124568.1 | - | 3 | 204 | |||
CR542132.1 | - | 2 | 73 | |||
L04483.1 | - | 3 | 111 | |||
X79563.2 | - | 3 | 110 |