Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000331.5 | NP_000322.2 | 4 | 673 | ||
NM_001178006.2 | NP_001171477.1 | 5 | 587 | |||
NM_199161.4 | NP_954630.1 | 4 | 526 | |||
GenBank mRNA | BC007022.1 | - | 4 | 487 | 4071612 | |
BC105796.1 | - | 5 | 552 | 4722684 | ||
BG562565.1 | - | 4 | 526 | |||
BQ691948.1 | - | 3 | 389 | |||
BX102362.1 | - | 4 | 593 | |||
CD102084.1 | - | 4 | 490 | |||
X51439.1 | - | 1 | 234 | |||
X51441.1 | - | 1 | 236 |
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X51442.1 | - | 1 | 236 |