Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021920.3 | NP_068739.1 | 2 | 187 | ||
XM_017018109.1 | XP_016873598.1 | 2 | 183 | |||
GenBank mRNA | HY019819.1 | - | 2 | 187 |