Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002987.2 | NP_002978.1 | 3, 4 | 319 | ||
XM_011523256.2 | XP_011521558.1 | 5, 6 | 690 | |||
XM_017023530.1 | XP_016879019.1 | 5, 6 | 693 | |||
GenBank mRNA | BC069107.1 | - | 3, 4 | 319 | 7216923 | |
BC112066.1 | - | 3, 4 | 285 | 8327534 | ||
BC112068.1 | - | 3, 4 | 288 | 8327536 | ||
D43767.1 | - | 2, 3 | 242 |