Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 12, 13 | 3197 | ||
NM_021574.2 | NP_067585.2 | 12, 13 | 3197 | |||
GenBank mRNA | AB209991.1 | - | 12, 13 | 2946 | ||
AF192533.1 | - | 1, 2 | 19 | |||
AK128501.1 | - | 4, 5 | 1254 | |||
AK303241.1 | - | 12, 13 | 1631 | |||
BC066122.1 | - | 12, 13 | 3028 | |||
BQ422888.1 | - | 3, 4 | 323 | |||
X02596.1 | - | 12, 13 | 3089 | |||
Y00661.1 | - | 12, 13 | 3197 |