Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003064.3 | NP_003055.1 | 3 | 335 | ||
GenBank mRNA | AF114471.1 | - | 3 | 314 | ||
AK312192.1 | - | 3 | 319 | |||
AX772818.1 | - | 3 | 335 | |||
BC020708.1 | - | 3 | 335 | 4733996 | ||
X04470.1 | - | 3 | 332 | |||
X04503.1 | - | 3 | 314 |