Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003087.2 | NP_003078.2 | 5 | 597 | ||
XM_011540105.2 | XP_011538407.1 | 5 | 592 | |||
GenBank mRNA | AF010126.1 | - | 5 | 441 | ||
AF017256.1 | - | 5 | 478 | |||
AF411524.1 | - | 5 | 441 | |||
AK223367.1 | - | 5 | 554 | |||
BC014098.2 | - | 5 | 500 | 4546444 |