Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003087.2 | NP_003078.2 | 2 | 313 | ||
XM_011540105.2 | XP_011538407.1 | 2 | 256 | |||
GenBank mRNA | AF010126.1 | - | 2 | 157 | ||
AF017256.1 | - | 2 | 194 | |||
AF411524.1 | - | 2 | 157 | |||
AK223367.1 | - | 2 | 270 | |||
AK312141.1 | - | 2 | 196 | |||
BC014098.2 | - | 2 | 216 | 4546444 | ||
BP346786.1 | - | 2 | 408 | |||
CR541788.1 | - | 2 | 146 | |||
CR541790.1 | - | 2 | 146 |