Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003278.2 | NP_003269.2 | 1, 2 | 209 | ||
GenBank mRNA | AK312093.1 | - | 1, 2 | 217 | ||
BC011024.1 | - | 1, 2 | 174 | 4042921 | ||
BI837909.1 | - | 1, 2 | 293 | |||
CD616729.1 | - | 1, 2 | 183 | |||
CR541981.1 | - | 1, 2 | 113 | |||
CR542009.1 | - | 1, 2 | 113 | |||
X64559.1 | - | 1, 2 | 206 |