Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003278.2 | NP_003269.2 | 1 | 190 | ||
XM_017007116.1 | XP_016862605.1 | 1 | 250 | |||
GenBank mRNA | AK312093.1 | - | 1 | 198 | ||
BC011024.1 | - | 1 | 155 | 4042921 | ||
BI837909.1 | - | 1 | 274 | |||
CD616729.1 | - | 1 | 164 | |||
CR541981.1 | - | 1 | 94 | |||
CR542009.1 | - | 1 | 94 | |||
X64559.1 | - | 1 | 187 |