Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SF1 SIRNA Assays ›
Gene Symbol:
SF1
Gene Name:
splicing factor 1
Gene Aliases:
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
Chromosome Location:
Chr.11: 64764604 - 64779043 on Build GRCh38
UniGene ID:
Hs.502829
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194850
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178030.1
NP_001171501.1
2
583
NM_001178031.1
NP_001171502.1
2
365
NM_004630.3
NP_004621.2
2
586
NM_201995.2
NP_973724.1
2
586
NM_201997.2
NP_973726.2
2
586
NM_201998.2
NP_973727.1
2
586
NR_033649.1
2
586
NR_033650.1
2
586
XM_011545244.1
XP_011543546.1
2
258
XM_011545245.1
XP_011543547.1
2
258
XM_011545246.1
XP_011543548.1
2
258
XM_011545247.1
XP_011543549.1
2
258
XM_011545248.1
XP_011543550.1
2
258
XM_017018244.1
XP_016873733.1
2
258
XM_017018245.1
XP_016873734.1
2
339
XM_017018246.1
XP_016873735.1
2
258
XM_017018247.1
XP_016873736.1
2
301
XM_017018248.1
XP_016873737.1
2
301
XM_017018249.1
XP_016873738.1
2
785
XM_017018250.1
XP_016873739.1
2
511
XM_017018251.1
XP_016873740.1
2
366
GenBank mRNA
AK293753.1
-
2
365
AK296237.1
-
2
284
AK301803.1
-
2
511
BC008080.2
-
2
282
2961666
BC008724.2
-
2
282
2961666
BC020217.1
-
2
485
4869292
BC034451.1
-
2
511
BC038446.1
-
2
606
5493676
BX095574.1
-
2
251
D26120.1
-
2
516
D26121.1
-
2
516
L49345.1
-
2
472
L49380.1
-
2
472
Y08765.1
-
2
176
Y08766.1
-
2
428
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
male sex determination
Leydig cell differentiation
negative regulation of smooth muscle cell proliferation
regulation of steroid biosynthetic process
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline