Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318949.1 | NP_001305878.1 | 5, 6 | 703 | ||
NM_001318950.1 | NP_001305879.1 | 5, 6 | 848 | |||
NM_001318951.1 | NP_001305880.1 | 4, 5 | 630 | |||
NM_003459.4 | NP_003450.2 | 4, 5 | 766 | |||
GenBank mRNA | AK313600.1 | - | 4, 5 | 721 | ||
BC028358.2 | - | 4, 5 | 754 | 4837595 | ||
U76010.1 | - | 4, 5 | 663 |