Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143993.1 | NP_001137465.1 | 3 | 776 | ||
NM_001143994.1 | NP_001137466.1 | 3 | 741 | |||
NM_003475.3 | NP_003466.1 | 3 | 741 | |||
XM_005253149.3 | XP_005253206.2 | 4 | 892 | |||
XM_005253150.1 | XP_005253207.1 | 3 | 803 | |||
XM_006718338.2 | XP_006718401.1 | 3 | 1144 | |||
XM_011520389.1 | XP_011518691.1 | 3 | 798 | |||
XM_017018360.1 | XP_016873849.1 | 4 | 892 | |||
XM_017018361.1 | XP_016873850.1 | 3 | 803 | |||
XM_017018362.1 | XP_016873851.1 | 3 | 803 | |||
GenBank mRNA | AL535761.3 | - | 3 | 771 | ||
BC106921.1 | - | 3 | 749 | 40032773 | ||
BC106922.1 | - | 3 | 749 | 40032778 | ||
M91083.1 | - | 3 | 749 |