Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300918.1 | NP_001287847.1 | 2 | 940 | ||
NM_001300919.1 | NP_001287848.1 | 2 | 940 | |||
NM_003483.4 | NP_003474.1 | 2 | 940 | |||
NM_003484.1 | NP_003475.1 | 2 | 940 | |||
XM_017019989.1 | XP_016875478.1 | 2 | 589 | |||
GenBank mRNA | AB209853.1 | - | 2 | 502 | ||
AF533651.1 | - | 1 | 18 | |||
AF533653.1 | - | 32 | ||||
AF533654.1 | - | 1 | 32 | |||
AI290477.1 | - | 1 | 18 | |||
AK311399.1 | - | 2 | 589 | |||
AY601861.1 | - | 2 | 940 | |||
AY601862.1 | - | 2 | 940 | |||
AY601863.1 | - | 2 | 940 | |||
AY601864.1 | - | 2 | 940 | |||
AY601865.1 | - | 2 | 940 | |||
AY601866.1 | - | 2 | 940 | |||
AY601867.1 | - | 2 | 940 | |||
U28749.1 | - | 2 | 940 | |||
X92518.1 | - | 2 | 1439 | |||
Z31595.1 | - | 2 | 941 |