Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003510.2 | NP_003501.1 | 1 | 46 | ||
GenBank mRNA | AK311941.1 | - | 1 | 79 | ||
BC034487.2 | - | 1 | 82 | 4825623 | ||
BC104198.1 | - | 1 | 46 | 40031339 | ||
BC104199.1 | - | 1 | 46 | 40031340 |