Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H4K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003541.2 | NP_003532.1 | 1 | 67 | ||
GenBank mRNA | AK312028.1 | - | 1 | 100 |
Gene Symbol: HIST1H4J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021968.3 | NP_068803.1 | 1 | 67 | ||
GenBank mRNA | AF086564.1 | - | 1 | 84 | ||
BC017361.2 | - | 1 | 86 | 4647914 | ||
BC148271.1 | - | 1 | 89 | 8860214 |