Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004711.4 | NP_004702.2 | 2 | 335 | ||
NM_145731.3 | NP_663783.1 | 2 | 335 | |||
NM_145738.2 | NP_663791.1 | 2 | 301 | |||
GenBank mRNA | AJ002303.1 | - | 2 | 301 | ||
AJ002304.1 | - | 2 | 273 | |||
AJ002305.1 | - | 2 | 273 | |||
AK289507.1 | - | 2 | 282 | |||
AK295863.1 | - | 2 | 293 | |||
BC000731.2 | - | 2 | 299 | 3508457 | ||
BI667990.1 | - | 2 | 359 | |||
BT007135.1 | - | 2 | 273 | |||
CR456590.1 | - | 2 | 283 |