Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005483.2 | NP_005474.2 | 3 | 230 | ||
XM_011527605.2 | XP_011525907.1 | 3 | 297 | |||
XM_011527606.2 | XP_011525908.1 | 3 | 297 | |||
XM_011527607.2 | XP_011525909.1 | 3 | 297 | |||
GenBank mRNA | BC041606.1 | - | 3 | 155 | ||
BC052620.1 | - | 3 | 229 | |||
BC067093.1 | - | 3 | 231 | 6339027 | ||
BQ889365.1 | - | 3 | 230 | |||
U20979.1 | - | 3 | 116 |