Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267551.1 | NP_001254480.1 | 4, 5 | 474 | ||
NM_001267552.1 | NP_001254481.1 | 4, 5 | 540 | |||
NM_007169.2 | NP_009100.2 | 4, 5 | 532 | |||
NM_148172.2 | NP_680477.1 | 4, 5 | 540 | |||
NM_148173.1 | NP_680478.1 | 4, 5 | 419 | |||
XM_006721418.3 | XP_006721481.2 | 4, 5 | 421 | |||
XM_017024016.1 | XP_016879505.1 | 2, 3 | 327 | |||
GenBank mRNA | AB029821.1 | - | 4, 5 | 532 | ||
AF044214.1 | - | 4, 5 | 505 | |||
AF113126.1 | - | 4, 5 | 509 | |||
AF176806.1 | - | 4, 5 | 482 | |||
AF176807.1 | - | 4, 5 | 427 | |||
AK302251.1 | - | 4, 5 | 545 | |||
BC000557.2 | - | 4, 5 | 516 | 3162576 | ||
BC050593.1 | - | 4, 5 | 517 | 6148983 | ||
BM011377.1 | - | 4, 5 | 402 | |||
CR457099.1 | - | 3, 4 | 352 |