Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 2 | 297 | ||
NR_073029.1 | 2 | 297 | ||||
XM_011539098.1 | XP_011537400.1 | 2 | 235 | |||
XM_011539099.1 | XP_011537401.1 | 2 | 235 | |||
GenBank mRNA | AF030162.1 | - | 2 | 249 | ||
AK223331.1 | - | 2 | 237 | |||
AK304883.1 | - | 2 | 232 | |||
AY442341.1 | - | 2 | 249 | |||
BC062707.1 | - | 2 | 301 | 4772587 | ||
BC066951.1 | - | 2 | 247 | 5264814 | ||
CD514420.1 | - | 2 | 173 |
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098044.1 | - | 3 | 396 | ||
AK295370.1 | - | 2 | 239 | |||
AK316490.1 | - | 2 | 239 | |||
AK316549.1 | - | 2 | 288 |