Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042618.1 | NP_001036083.1 | 11 | 1066 | ||
NM_005484.3 | NP_005475.2 | 11 | 1105 | |||
XM_005267247.3 | XP_005267304.1 | 11 | 1105 | |||
XM_017020912.1 | XP_016876401.1 | 11 | 1639 | |||
GenBank mRNA | AF085734.1 | - | 11 | 1102 | ||
AJ236876.2 | - | 10 | 1016 | |||
AJ236912.1 | - | 11 | 1040 | |||
AK001980.1 | - | 11 | 1082 | |||
AK290079.1 | - | 11 | 1045 | |||
AK300971.1 | - | 9 | 823 |