Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293634.1 | NP_001280563.1 | 11 | 2327 | ||
NM_021008.3 | NP_066288.2 | 12 | 2552 | |||
GenBank mRNA | AB209831.1 | - | 10 | 1453 | ||
AF007165.1 | - | 12 | 1700 | |||
AF049459.1 | - | 12 | 1895 | |||
AF049460.1 | - | 13 | 2159 | |||
AK289873.1 | - | 12 | 1778 | |||
AK291383.1 | - | 12 | 2333 | |||
BC053322.1 | - | 12 | 1960 | 6584290 | ||
BQ267366.1 | - | 1 | 43 |