Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006551.3 | NP_006542.1 | 2 | 234 | ||
GenBank mRNA | AJ224172.1 | - | 2 | 260 | ||
AL708956.1 | - | 2 | 259 | |||
BC069290.1 | - | 2 | 228 | 7262705 | ||
BC104838.1 | - | 2 | 204 | 8143841 | ||
BC104840.1 | - | 2 | 203 | 8143843 |