Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003931.3 | NP_001003931.3 | 4 | 753 | ||
NM_005485.5 | NP_005476.4 | 4 | 758 | |||
XM_005264779.4 | XP_005264836.2 | 5 | 898 | |||
XM_017005490.1 | XP_016860979.1 | 5 | 885 | |||
GenBank mRNA | AB208803.1 | - | 4 | 638 | ||
AF083068.1 | - | 4 | 477 | |||
AL050034.1 | - | 2 | 126 | |||
AY126341.1 | - | 4 | 588 | |||
BC014260.2 | - | 3 | 359 | 4763951 | ||
BI554046.1 | - | 4 | 692 | |||
Y16836.1 | - | 4 | 712 |