Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008220.1 | NP_001008221.1 | 4 | 915 | ||
NM_006650.3 | NP_006641.1 | 5 | 965 | |||
XM_005265798.1 | XP_005265855.1 | 4 | 843 | |||
XM_005265799.1 | XP_005265856.1 | 4 | 953 | |||
XM_011534419.1 | XP_011532721.1 | 4 | 1231 | |||
XM_017008964.1 | XP_016864453.1 | 4 | 973 | |||
GenBank mRNA | AK124837.1 | - | 2 | 66 |