Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014449.2 | NP_001014449.1 | 10 | 1694 | ||
NM_001014451.2 | NP_001014451.1 | 11 | 1704 | |||
NM_001257172.1 | NP_001244101.1 | 11 | 1629 | |||
NM_001257173.1 | NP_001244102.1 | 11 | 1786 | |||
NM_001257174.1 | NP_001244103.1 | 10 | 1740 | |||
NM_001257175.1 | NP_001244104.1 | 9 | 1601 | |||
NM_007242.5 | NP_009173.1 | 12 | 1797 | |||
XM_006721127.2 | XP_006721190.1 | 11 | 1719 | |||
XM_011522832.2 | XP_011521134.1 | 13 | 1853 | |||
XM_011522833.2 | XP_011521135.1 | 12 | 1760 | |||
XM_011522834.2 | XP_011521136.1 | 12 | 1742 | |||
XM_017022890.1 | XP_016878379.1 | 12 | 1835 | |||
GenBank mRNA | AF353720.1 | - | 10 | 1741 | ||
AK027378.1 | - | 12 | 1762 | |||
AK223115.1 | - | 12 | 1768 | |||
AK296868.1 | - | 9 | 1598 | |||
AK316346.1 | - | 10 | 1675 | |||
AL136639.1 | - | 11 | 1673 | |||
BC003626.2 | - | 12 | 1761 | 3030038 | ||
BC010008.2 | - | 10 | 1646 | 4298615 | ||
BC110805.1 | - | 11 | 1568 |