Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015193.4 | NP_056008.1 | 1 | 1769 | ||
GenBank mRNA | AF193421.1 | - | 1 | 1757 | ||
BC012321.1 | - | 1 | 1773 | 4555433 | ||
BC075802.1 | - | 1 | 1761 | 5198843 | ||
D87468.1 | - | 1 | 1761 |