Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015414.3 | NP_056229.2 | 3 | 511 | ||
NM_033643.2 | NP_378669.1 | 4 | 407 | |||
GenBank mRNA | AF077043.1 | - | 3 | 483 | ||
AL050273.1 | - | 4 | 373 | |||
BC003052.1 | - | 3 | 490 | 3546966 | ||
BC004971.1 | - | 4 | 362 | 2957859 | ||
BC091508.1 | - | 4 | 390 | 30417606 | ||
BM015858.1 | - | 3 | 511 | |||
BM841746.1 | - | 4 | 407 |
Gene Symbol: DHX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL359945.1 | - | 2266 | |||
CR936655.1 | - | 2266 |
Gene Symbol: XPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC050680.1 | - | 2486 |